Gold 99.99% (trace metals basis), shot

Supplier: Thermo Scientific Chemicals

43714-0050 437140010
TS43714-0050EA 1364 USD
TS43714-0050 TS437140010
Gold 99.99% (trace metals basis), shot
Gold
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800 °C (760 mmHg)
Melting Pt: 1065 °C
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5

Order Now

Specification Test Results

Appearance Golden Beads or pellets
Total trace metal impurities ≤100 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR