Tetrabutylammonium hydrogen sulfate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tetrabutylammonium bisulphate, Tetrabutylammonium hydrogensulphate

16838-0250 16838-1000 16838-2500 AC16838-0010
TS16838-0250EA 67 USD
TS16838-0250 TS16838-1000 TS16838-2500 TS16838-0010
Tetrabutylammonium hydrogen sulfate 98%
Tetrabutylammonium hydrogen sulphate
Formula: (C₄H₉)₄NHSO₄
MW: 339.54 g/mol
Melting Pt: 168…172 °C
MDL Number: MFCD00011637
CAS Number: 32503-27-8
EINECS: 251-068-5

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 168 to 173 °C
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR