Copper(II) sulfate pentahydrate 98+% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cupric sulphate pentahydrate, Copper sulphate pentahydrate

42361-0030 AC42361-0100 AC42361-5000
TS42361-0030EA 477 USD
TS42361-0030 TS42361-0100 TS42361-5000
Copper(II) sulfate pentahydrate 98+% ACS
Copper(II) sulphate pentahydrate
Formula: CuSO₄·5H₂O
MW: 249.69 g/mol
Melting Pt: 110 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149681
CAS Number: 7758-99-8
EINECS: 231-847-6
UN: 3077
ADR: 9,III
REACH: 01-2119520566-40
Merck Index: 13,02682

Order Now

Specification Test Results

Appearance Blue Crystals
Titration Iodometric 98.0 to 102.0 %
Chloride (Cl) ≤10 ppm
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤30 ppm
Nickel (Ni) ≤50 ppm
Potassium (K) ≤100 ppm
Sodium (Na) ≤200 ppm
Insoluble matter ≤0.005 %
Nitrogen compounds (as N) ≤0.002 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR