1-Butanesulfonic acid sodium salt 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Sodium 1-butanesulphonate, Sodium butane-1-sulphonate, Sodium 1-butansulphonate, Butane-1-sulphonic acid sodium salt
20610-0250
AC20610-1000
TS20610-0250EA
192
USD
TS20610-0250
TS20610-1000
1-Butanesulfonic acid sodium salt 98%
1-Butanesulphonic acid sodium salt
Formula:
C₄H₉NaO₃S MW: 160.17 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007540 CAS Number: 2386-54-1 EINECS: 219-201-1 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤2 % (1 g, 105°C, 2 h) |
Solubility | (10 % in water) Clear to slightly hazy colorless |
Titration after Ion exchange | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...