Ammonium thiocyanate 98% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Thiocyanic acid ammonium salt, Ammonium rhodanide, Ammonium sulphocyanate
42341-5000
AC42341-0030
TS42341-5000EA
165
USD
TS42341-5000
TS42341-0030
Ammonium thiocyanate 98% ACS
Ammonium thiocyanate
Formula:
NH₄SCN MW: 76.12 g/mol Melting Pt: 150 Density: 1.3 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011428 CAS Number: 1762-95-4 EINECS: 217-175-6 Merck Index: 13,00564 |
Specification Test Results
Appearance | Colorless to white Crystals |
Titration Argentimetric | ≥97.5 % |
Residue after ignition | ≤0.025 % |
pH | 4.5 to 6.0 (5 % at 25.0°C) |
Chloride (Cl) | ≤0.005 % |
Sulfate (SO4) | ≤0.005 % |
Iron (Fe) | ≤3 ppm |
Heavy metals (as Pb) | ≤5 ppm |
Insoluble matter | ≤0.005 % |
Iodine consuming subst. | ≤0.004 meq/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...