Potassium peroxodisulphate 99+% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
di-Potassium peroxodisulphate, Peroxydisulphuric acid dipotassium salt, Dipotassium peroxodisulphate, Potassium persulphate
42418-5000
AC42418-0025
TS42418-5000EA
107
USD
TS42418-5000
TS42418-0025
Potassium peroxodisulphate 99+% ACS
Potassium peroxodisulphate
Formula:
K₂S₂O₈ MW: 270.32 g/mol Melting Pt: 100 Storage Temperature: Ambient |
MDL Number:
MFCD00011386 CAS Number: 7727-21-1 EINECS: 231-781-8 UN: 1492 ADR: 5.1 III Merck Index: 13,07741 |
Specification Test Results
Appearance | White Crystallline powder or crystals |
Titration with KMnO4 | ≥99.0 % |
Chlorine compounds (as Cl) | ≤0.001 % |
Iron (Fe) | ≤5 ppm |
Manganese (Mn) | ≤2 ppm |
Heavy metals (as Pb) | ≤0.001 % |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...