Potassium disulphate 98% for analysis

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Potassium disulphate (Potassium pyrosulphate), Dipotassium disulphate, Potassium pyrosulphate

20791-0025 20791-5000
TS20791-0025EA 718 USD
TS20791-0025 TS20791-5000
Potassium disulphate 98% for analysis
Potassium disulphate
Formula: K₂O₇S₂
Melting Pt: 325 °C
Density: 2.51 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011385
CAS Number: 7790-62-7
EINECS: 232-216-8
UN: 2923
ADR: 8,I
Merck Index: 13,07749

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration with NaOH ≥97.5 %
Chloride (Cl) ≤10 ppm
Phosphate (PO4) ≤10 ppm
Arsenic (As) ≤5 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm
Ca and Mg ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR