Ammonium dihydrogen phosphate 99.999% (trace metals basis)

Supplier: Thermo Scientific Chemicals

43679-1000 AC43679-0250 AC43679-5000
TS43679-1000EA 162 USD
TS43679-1000 TS43679-0250 TS43679-5000
Ammonium dihydrogen phosphate 99.999% (trace metals basis)
Ammonium dihydrogen phosphate
Formula: NH₄H₂PO₄
Melting Pt: 190 °C
Storage Temperature: Ambient
MDL Number: MFCD00003396
CAS Number: 7722-76-1

Order Now

Specification Test Results

Appearance Colorless to white Crystals
Total trace metal impurities ≤10 ppm
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR