Dipotassium hydrogen orthophosphate trihydrate, Extra Pure

Supplier: Thermo Scientific Chemicals

44773-2500 AC44773-0010
TS44773-2500EA 65 USD
TS44773-2500 TS44773-0010
Dipotassium hydrogen orthophosphate trihydrate, Extra Pure
Dipotassium hydrogen orthophosphate trihydrate
Formula: K₂HPO₄·3H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149926
CAS Number: 16788-57-1

Order Now

Specification Test Results

Appearance White Crystals
pH 8.5 to 9.6 (5 % solution)
Titration with HClO4 ≥98.5 %
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤100 ppm
Arsenic (As) ≤1 ppm
Iron (Fe) ≤30 ppm
Sodium (Na) ≤0.1 %
Heavy metals (as Pb) ≤10 ppm
Nitrogen compounds ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR