Pyrogallol ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Benzene-1,2,3-triol, 1,2,3-Trihydroxybenzene
41862-1000
41862-0025
41862-5000
TS41862-1000EA
210
USD
TS41862-1000
TS41862-0025
TS41862-5000
Pyrogallol ACS
Pyrogallol
Formula:
C₆H₆O₃ Boiling Pt: 309 °C (1013 hPa) Melting Pt: 131…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002192 CAS Number: 87-66-1 EINECS: 201-762-9 UN: 2811 ADR: 6.1,III Merck Index: 13,08090 |
|
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 131.0 to 135.0 °C |
Residue after ignition | ≤0.005 % |
Chloride (Cl) | ≤10 ppm |
Sulfate (SO4) | ≤50 ppm |
Iron (Fe) | ≤10 ppm |
Heavy metals (as Pb) | ≤5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...