Diethylene glycol 99%, Extra Pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,2'-Oxybis(ethan-1-ol), Digol, 2,2'-Oxybisethanol

12116-0025 12116-0100 12116-0010
TS12116-0025EA 116 USD
TS12116-0025 TS12116-0100 TS12116-0010
Diethylene glycol 99%, Extra Pure
Diethylene glycol
Formula: C₄H₁₀O₃
MW: 106.12 g/mol
Boiling Pt: 245 °C (760 mmHg)
Melting Pt: –10 °C
Density: 1.11 g/cm³
Flash Pt: 143 °C
Storage Temperature: Ambient
MDL Number: MFCD00002882
CAS Number: 111-46-6
EINECS: 203-872-2
Merck Index: 13,03146
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f023bd53ecff654f6d6d398b3ee2a1bac87d56f&vid=25d8943f9c247b4203b1e1bcba59bf77dfdd9158&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear viscous liquid
Infrared spectrum Conforms
Refractive index 1.4450 to 1.4480 (20°C, 589 nm)
Water ≤0.2 %
GC ≥98.5 %
Color scale ≤20 APHA
Ash ≤0.01 %
Acidity (CH3COOH) ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR