2-Butoxyethanol 99%, Extra Pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethylene glycol butyl ether, Butyl cellosolve, Ethylene glycol monobutyl ether

15433-0010 15433-0100 154330025 154330250
TS15433-0010EA 55 USD
TS15433-0010 TS15433-0100 TS154330025 TS154330250
2-Butoxyethanol 99%, Extra Pure
2-Butoxyethanol
Formula: CH₃(CH₂)₃OCH₂CH₂OH
MW: 118.18 g/mol
Boiling Pt: 171 °C (760 mmHg)
Melting Pt: –70 °C
Density: 0.901 g/cm³
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00002884
CAS Number: 111-76-2
EINECS: 203-905-0
Merck Index: 13,01558
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba5a9d2c527d0c167d2248d1936e2d1bd4aeab7a&vid=75260b2608ddc5eb29642178ab193fbcaf54198d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4180 to 1.4220 (20 °C, 589 nm)
Water ≤0.1 % (Coulometric)
Density (20°C) 0.899 to 0.903 kg/l
GC ≥98.5 %
Color scale ≤10 APHA
Residue after evaporation ≤0.02 %
Acidity (CH3COOH) ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR