Dimethyl sulfoxide-[D₆] 99.9 atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals

16629-0100 16629-0500 16629-1000 166290101 166290250
TS16629-0100EA 56.38 USD
TS16629-0100 TS16629-0500 TS16629-1000 TS166290101 TS166290250
Dimethyl sulfoxide-[D₆] 99.9 atom % D for NMR spectroscopy
Dimethyl sulphoxide-[D₆]
Formula: D₃CS(O)CD₃
Boiling Pt: 189 °C (5 mmHg)
Melting Pt: 20.2 °C
Density: 1.19 g/cm³
Flash Pt: 88 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002090
CAS Number: 2206-27-1
EINECS: 218-617-0

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.03 % (K.F.)
D-Enrichment ≥99.85 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR