Deuterium oxide 99.8 atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals

16630-0010 166300250 166301000 166305000
TS16630-0010EA 1607.7 USD
TS16630-0010 TS16630-0100 TS166300250 TS166301000 TS166305000
Deuterium oxide 99.8 atom % D for NMR spectroscopy
Deuterium oxide
Formula: D₂O
Boiling Pt: 101.3 °C (1013 hPa)
Melting Pt: 3.8 °C
Density: 1.105 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00044636
CAS Number: 7789-20-0
EINECS: 232-148-9
Merck Index: 12,02984

Order Now

Specification Test Results

Appearance Clear colorless Liquid
D-Enrichment ≥99.75 %
Melting point 69 to 77 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR