Dimethyl sulphoxide 99.8+% for peptide synthesis
Supplier: Thermo Scientific Chemicals
Synonyms:
Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO
35484-5000
354840025
TS35484-5000EA
121
USD
TS35484-5000
TS354840025
Dimethyl sulphoxide 99.8+% for peptide synthesis
Dimethyl sulphoxide
Formula:
(CH₃)₂SO Boiling Pt: 189 °C (760 mmHg) Melting Pt: 18.4 °C Density: 1.1 g/cm³ Flash Pt: 87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002089 CAS Number: 67-68-5 EINECS: 200-664-3 Merck Index: 13,03285 |
|
Specification Test Results
Appearance | Clear colorless Liquid |
Water | ≤0.005 % (K.F.) |
GC | ≥99.80 % |
Residue after evaporation | ≤0.001 % |
Free amine | ≤0.0002 % |
Acidity (CH3COOH) | ≤0.003 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...