Dimethyl sulphoxide 99.7%, pure

Supplier: Thermo Scientific Chemicals

Synonyms: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO

12779-0010 12779-0050 12779-0025 127790250 127791000
TS12779-0010EA 116 USD
TS12779-0010 TS12779-0050 TS12779-0025 TS127790250 TS127791000
Dimethyl sulphoxide 99.7%, pure
Dimethyl sulphoxide
Formula: (CH₃)₂SO
Boiling Pt: 189 °C (760 mmHg)
Melting Pt: 18.4 °C
Density: 1.1 g/cm³
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4770 to 1.4800 (20 °C, 589 nm)
Water ≤0.2 %
GC ≥99.6 %
Color scale ≤20 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR