Cobalt(II) nitrate hexahydrate 99%, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cobalt dinitrate hexahydrate, Cobaltous nitrate hexahydrate

21309-0050 21309-1000 AC21309-0010 AC21309-5000
TS21309-0050EA 808 USD
TS21309-0050 TS21309-1000 TS21309-0010 TS21309-5000
Cobalt(II) nitrate hexahydrate 99%, pure
Cobalt(II) nitrate hexahydrate
Formula: Co(NO₃)₂·6H₂O
MW: 291.03 g/mol
Melting Pt: 55 °C
Storage Temperature: Ambient
MDL Number: MFCD00149647
CAS Number: 10026-22-9
EINECS: 233-402-1
UN: 1477
ADR: 5.1,II
Merck Index: 13,02469

Order Now

Specification Test Results

Appearance Pink to red to brown Adhering crystalline powder or crystals or chips
Titration Complexometric ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR