Ammonium cerium(IV) nitrate 99% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
42354-0025
42354-1000
42354-5000
AC42354-0250
AC42354-0500
TS42354-0025EA
920
USD
TS42354-0025
TS42354-1000
TS42354-5000
TS42354-0250
TS42354-0500
Ammonium cerium(IV) nitrate 99% ACS
Ammonium cerium(IV) nitrate
Formula:
(NH₄)₂Ce(NO₃)₆ Melting Pt: 107…108 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151121 CAS Number: 16774-21-3 EINECS: 240-827-6 UN: 3085 ADR: 5.1,II Merck Index: 13,00509 |
Specification Test Results
Appearance | Orange Crystalline powder or crystals |
Titration | ≥98.5 % |
Chloride (Cl) | ≤0.01 % |
Phosphate (PO4) | ≤0.02 % |
Iron (Fe) | ≤0.005 % |
Insoluble matter | ≤0.05 % (in dilute sulfuric acid) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...