Bis-trispropane 99%
Supplier: Thermo Scientific Chemicals
20264-1000
20264-2500
TS20264-1000EA
188
USD
TS20264-1000
TS20264-2500
Bis-trispropane 99%
Bis-trispropane
Formula:
C₁₁H₂₆N₂O₆ Melting Pt: 164…165 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004689 CAS Number: 64431-96-5 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 161 to 168 °C |
Loss on drying | ≤1 % |
Solubility | (0.1 M in water) Clear colorless |
UV | at 400 nm A: ≤0.05 |
Titration with HClO4 | 98.5 to 101.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...