Diethylamine 99.5%, Extra Pure, redistilled

Supplier: Thermo Scientific Chemicals
Danger

37837-0010 37837-1000
TS37837-0010EA 387 USD
TS37837-0010 TS37837-1000
Diethylamine 99.5%, Extra Pure, redistilled
Diethylamine
Formula: (C₂H₅)₂NH
MW: 73.14 g/mol
Boiling Pt: 55 °C (760 mmHg)
Melting Pt: –50 °C
Density: 0.71 g/cm³
Flash Pt: –23 °C
Storage Temperature: Ambient
MDL Number: MFCD00009032
CAS Number: 109-89-7
EINECS: 203-716-3
UN: 1154
ADR: 3,II
Merck Index: 13,03138
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c054fa528239af221505ef08b7c0d2503c9d1d7&vid=e1e59ef071f0c580f9eb9716f4801d314ff824c3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3840 to 1.3860 (20°C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥99.4 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR