(3-Aminopropyl)triethoxysilane 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Aminopropyltriethoxysilane, 3-(Triethoxysilyl)propylamine, Geniosil GF 93, Triethoxy(3-aminopropyl)silane

15108-5000 AC15108-0025 AC15108-0050 AC15108-1000
TS15108-5000EA 325 USD
TS15108-5000 TS15108-0025 TS15108-0050 TS15108-1000
(3-Aminopropyl)triethoxysilane 99%
(3-Aminopropyl)triethoxysilane
Formula: H₂NCH₂CH₂CH₂Si(OCH₂CH₃)₃
MW: 221.37 g/mol
Boiling Pt: 217 °C (760 mmHg)
Melting Pt: –70 °C
Density: 0.942 g/cm³
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00008207
CAS Number: 919-30-2
EINECS: 213-048-4
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4200 to 1.4220 (20°C, 589 nm)
GC ≥98.5 %
Color scale ≤25 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR