Malachite green oxalate, pure, certified

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Malachite green hemioxalate, N,N,N',N'-Tetramethyl-4,4'-diaminotriphenylcarbenium oxalate, Bis (N-{4-[[4-(dimethylamino)phenyl](phenyl)methylene]cyclohexa-2,5-dien-1-ylidene}-N-methylmethanaminium) trioxalate, Basic Green 4, N-[4-[[4-(Dimethylamino)phenyl]phenylmethylene]-2,5-cyclohexadien-1-ylidene]-N-methyl-methanaminium, ethanedioate, ethanedioate(2:2:1)

41349-1000 41349-0250
TS41349-1000EA 144 USD
TS41349-1000 TS41349-0250
Malachite green oxalate, pure, certified
Malachite green oxalate
Formula: C₅₂H₅₄N₄O₁₂
MW: 927.02 g/mol
Melting Pt: 144…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00011766
CAS Number: 2437-29-8
EINECS: 219-441-7
UN: 2811
ADR: 6.1,III
Merck Index: 13,05722
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2dffa05d1d66e7e32371284ada0fcf1a0faa7d0&vid=950afba5ed94b79530cd91b1261e3c2223d2aecd&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7723ac34e7273345d2b069c0836679df388eaaf7&vid=ec3b640e950ddc753e47d460407fdd3277490d06&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Green to dark green Crystalline powder or crystals
Infrared spectrum Conforms
Dye content ≥90 %
UV Lambda max 614 to 620 nm (in water)
Absorption Ratio P-15/P+15 1.00 to 1.15
Visual transition interval from pH 11.6 (blue-green) to pH 14.0 (colorless)
BSC Certification Passes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR