Ammonium iodide ACS

Supplier: Thermo Scientific Chemicals
Warning

19321-0010 AC19321-0500 AC19321-2500
TS19321-0010EA 567 USD
TS19321-0010 TS19321-0500 TS19321-2500
Ammonium iodide ACS
Ammonium iodide
Formula: NH₄I
MW: 144.94 g/mol
Melting Pt: 551 (subl.)
Density: 2.51 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011424
CAS Number: 12027-06-4
Merck Index: 13,00531

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals (May turn yellow on storage)
Residue after ignition ≤0.05 %
Titration ≥99.0 %
Chloride and Bromide ≤0.005 % (as Cl)
Phosphate (PO4) ≤0.001 %
Sulfate (SO4) ≤0.05 %
Barium (Ba) ≤0.002 %
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤0.001 %
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR