Lead(II) iodide 99%

Supplier: Thermo Scientific Chemicals
Danger

19886-2500 AC19886-0500
TS19886-2500EA 248 USD
TS19886-2500 TS19886-0500
Lead(II) iodide 99%
Lead(II) iodide
Formula: I₂Pb
Boiling Pt: 954 °C (1013 hPa)
Melting Pt: 402 °C
Density: 6.16 g/cm³
MDL Number: MFCD00011163
CAS Number: 10101-63-0
EINECS: 233-256-9
UN: 2291
ADR: 6.1,III
Merck Index: 12,05430

Order Now

Specification Test Results

Appearance Yellow to orange Powder
Titration Argentometric ≥98.5 %
Loss on drying ≤0.1 %
Chloride (Cl) ≤100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR