Phenylhydrazinium chloride 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Phenylhydrazine hydrochloride, Phenylhydrazinium chloride

15155-1000 15155-5000
TS15155-1000EA 166 USD
TS15155-1000 TS15155-5000
Phenylhydrazinium chloride 99+%
Phenylhydrazinium chloride
Formula: C₆H₉ClN₂
Melting Pt: 250…254 °C
Storage Temperature: Ambient
MDL Number: MFCD00012924
CAS Number: 59-88-1
EINECS: 200-444-7
UN: 2811
ADR: 6.1,II
Merck Index: 12,07447
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e16d43c3611e20636f31c95d3974860d83c8913f&vid=c26f549d3aed1b69060ac1042fd1114ca4edb2cb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to light yellow to pink-beige Crystalline powder
Infrared spectrum Conforms
Solubility (5% in H2O) Clear colorless to light yellow solution
HPLC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR