Pentafluoropropionic anhydride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Perfluoropropanoic anhydride, Pentafluoropropanoic anhydride, PFPA

41693-0250 41693-0050 41693-1000
TS41693-0250EA 165 USD
TS41693-0250 TS41693-0050 TS41693-1000
Pentafluoropropionic anhydride 98%
Pentafluoropropionic anhydride
Formula: C₆F₁₀O₃
Boiling Pt: 69 °C
Density: 1.57 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000429
CAS Number: 356-42-3
EINECS: 206-604-2
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR