Difluoroacetic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

16212-0500 162120025 162120100
TS16212-0500EA 438 USD
TS16212-0500 TS162120025 TS162120100
Difluoroacetic acid 98%
Difluoroacetic acid
Formula: C₂H₂F₂O₂
MW: 96.03 g/mol
Boiling Pt: 132…134 °C (760 mmHg)
Melting Pt: –1 °C
Density: 1.52 g/cm³
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00004220
CAS Number: 381-73-7
EINECS: 206-839-0
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.3420 to 1.3450 (20°C, 589 nm)
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR