Cesium fluoride 99% for analysis

Supplier: Thermo Scientific Chemicals
Danger

18951-5000 AC18951-0250 AC18951-1000
TS18951-5000EA 516 USD
TS18951-5000 TS18951-0250 TS18951-1000
Cesium fluoride 99% for analysis
Cesium fluoride
Formula: CsF
MW: 151.9
Boiling Pt: 1251
Melting Pt: 680
Density: 4.627
Storage Temperature: Ambient
MDL Number: MFCD00010960
CAS Number: 13400-13-0
EINECS: 236-487-3
UN: 3288
ADR: 6.1 II

Order Now

Specification Test Results

Appearance White Crystalline powder
Assay ≥98.5 %
Chloride (Cl) ≤200 ppm
Sulfate (SO4) ≤300 ppm
Aluminium (Al) ≤10 ppm
Barium (Ba) ≤10 ppm
Calcium (Ca) ≤50 ppm
Lithium (Li) ≤5 ppm
Magnesium (Mg) ≤50 ppm
Potassium (K) ≤300 ppm
Rubidium (Rb) ≤200 ppm
Sodium (Na) ≤100 ppm
Strontium (Sr) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR