Thioacetamide 99+% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ethanethioamide
42453-1000
42453-5000
TS42453-1000EA
263
USD
TS42453-1000
TS42453-5000
Thioacetamide 99+% ACS
Thioacetamide
Formula:
CH₃CSNH₂ MW: 75.13 g/mol Melting Pt: 112…114 °C Density: 1.37 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00008070 CAS Number: 62-55-5 EINECS: 200-541-4 Merck Index: 13,09391 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 111 to 114 °C |
Titration Argentometric | ≥99.0 % |
Residue after ignition | ≤0.05 % |
Clarity of solution | (2% soln.) Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...