Sodium salicylate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium 2-hydroxybenzoate, 2-Hydroxybenzoic acid sodium salt, Salicylic acid sodium salt

22098-0010 22098-0025 22098-2500 22098-0500
TS22098-0010EA 381 USD
TS22098-0010 TS22098-0025 TS22098-2500 TS22098-0500
Sodium salicylate 97%
Sodium salicylate
Formula: HOC₆H₄COONa
MW: 160.10 g/mol
Boiling Pt: 30 °C (760 mmHg)
Melting Pt: 200
Density: 0.64 g/cm³
Flash Pt: –28 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002440
CAS Number: 54-21-7
EINECS: 203-694-5
UN: 1108
ADR: 3 I
Merck Index: 12,08819

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3704 to 1.3724 (20°C, 589 nm)
GC ≥96.0 %
Titration with HClO4 99.0 to 101.0 % (on dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR