(±)-α-Methoxyphenylacetic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
DL-α-Methoxyphenylacetic acid, 2-Methoxy-2-phenylacetic acid, α-Methoxyphenylacetic acid
41387-0050
41387-0250
41387-1000
41387-0010
TS41387-0050EA
139
USD
TS41387-0050
TS41387-0250
TS41387-1000
TS41387-0010
(±)-α-Methoxyphenylacetic acid 99%
(±)-α-Methoxyphenylacetic acid
Formula:
C₉H₁₀O₃ MW: 166.18 g/mol Melting Pt: 68…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064217 CAS Number: 7021-09-2 EINECS: 230-300-9 |
Specification Test Results
Appearance | Off-white to light beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 68 to 72 °C |
Titration with NaOH | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...