Ammonium formate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Formic acid ammonium salt

40115-0010 40115-0050 40115-2500
TS40115-0010EA 159 USD
TS40115-0010 TS40115-0050 TS40115-2500
Ammonium formate 99%
Ammonium formate
Formula: HCO₂NH₄
MW: 63.06 g/mol
Melting Pt: 115…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00013103
CAS Number: 540-69-2
EINECS: 208-753-9
Merck Index: 13,00525

Order Now

Specification Test Results

Appearance White Powder or crystals
Infrared spectrum Conforms
Titration excess NaOH ≥98.5 %
Chloride (Cl) ≤0.005 %
Sulfate (SO4) ≤0.01 %
Iron (Fe) ≤0.002 %
Heavy metals (as Pb) ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR