Ammonium formate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Formic acid ammonium salt

16861-0010 AC16861-0051 AC16861-5000
TS16861-0010EA 124 USD
TS16861-0010 TS16861-0051 TS16861-5000
Ammonium formate 97%
Ammonium formate
Formula: HCO₂NH₄
MW: 63.06 g/mol
Melting Pt: 115…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00013103
CAS Number: 540-69-2
EINECS: 208-753-9
Merck Index: 13,00525

Order Now

Specification Test Results

Appearance White Powder or crystals
Infrared spectrum Conforms
Titration with HClO4 96.0 to 104.0 %
HPLC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR