Oxalyl dichloride 2.0 M in dichloromethane, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Oxalyl chloride

36918-1000 369188000
TS36918-1000EA 72 USD
TS36918-1000 TS369188000
Oxalyl dichloride 2.0 M in dichloromethane, AcroSeal™
Oxalyl dichloride
Formula: C₂Cl₂O₂
MW: 126.93 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00000704
CAS Number: 79-37-8
EINECS: 201-200-2
UN: 2922
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Solution
Titration Argentometric 1.9 to 2.1 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR