Ammonium hydrogen carbonate 99% for analysis

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ammonium bicarbonate

39321-2500 AC39321-0010 AC39321-0050
TS39321-2500EA 50 USD
TS39321-2500 TS39321-0010 TS39321-0050
Ammonium hydrogen carbonate 99% for analysis
Ammonium hydrogen carbonate
Formula: NH₄HCO₃
MW: 79.06 g/mol
Melting Pt: 35 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012138
CAS Number: 1066-33-7
Merck Index: 13,00497

Order Now

Specification Test Results

Appearance White Crystalline powder
Sulfated ash ≤0.05 %
Titration with NaOH ≥98.5 %
Chloride (Cl) ≤5 ppm
Phosphate (PO4) ≤5 ppm
Calcium (Ca) ≤10 ppm
Copper (Cu) ≤5 ppm
Iron (Fe) ≤5 ppm
Potassium (K) ≤10 ppm
Sodium (Na) ≤20 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR