1,1'-Carbonyldiimidazole 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Di(1H-imidazol-1-yl)methanone, CDI Crosslinker, 1,1'-Carbonylbis(imidazole), 1,1'-Carbonylbis-1H-imidazole, N,N'-Carbonyldiimidazole, CDI
15181-0250
15181-1000
15181-0100
15181-5000
TS15181-0250EA
120
USD
TS15181-0250
TS15181-1000
TS15181-0100
TS15181-5000
1,1'-Carbonyldiimidazole 97%
1,1'-Carbonyldiimidazole
Formula:
C₇H₆N₄O MW: 162.15 g/mol Melting Pt: 118…120 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005286 CAS Number: 530-62-1 EINECS: 208-488-9 UN: 3259 ADR: 8 II Merck Index: 14,01819 |
Specification Test Results
Appearance | White to cream Crystalline powder |
Infrared spectrum | Conforms |
Assay | ≥96.0 % |
Water | ≤0.5 % |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...