1-Ethyl-2-pyrrolidone 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Ethyl-2-pyrrolidinone, N-Ethyl-2-pyrrolidone, 1-Ethylpyrrolidin-2-one

16393-5000 16393-0250 16393-1000
TS16393-5000EA 408 USD
TS16393-5000 TS16393-0250 TS16393-1000
1-Ethyl-2-pyrrolidone 98%
1-Ethyl-2-pyrrolidone
Formula: C₆H₁₁NO
MW: 113.16 g/mol
Boiling Pt: 212.5 °C (760 mmHg)
Melting Pt: –75 °C
Density: 0.998 g/cm³
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00003199
CAS Number: 2687-91-4
EINECS: 220-250-6

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4640 to 1.4680 (20°C, 589 nm)
GC ≥97.5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR