Choline chloride 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
2-hydroxy-N,N,N-trimethylethanaminium chloride
11029-0050
11029-5000
TS11029-0050EA
410
USD
TS11029-0050
TS11029-5000
Choline chloride 99%
Choline chloride
Formula:
HOH₂CCH₂N(CH₃)₃Cl MW: 139.62 g/mol Melting Pt: 298…304 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011721 CAS Number: 67-48-1 EINECS: 200-655-4 Merck Index: 13,02226 |
|
Specification Test Results
Appearance | White Adhering crystals |
Infrared spectrum | Conforms |
Titration Argentometric | 98.5 to 101.5 % (on dry substance) |
Water | ≤2 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...