Chloroacetamide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloroacetamide

14841-0050 14841-0010 14841-5000
TS14841-0050EA 37 USD
TS14841-0050 TS14841-0010 TS14841-5000
Chloroacetamide 98%
Chloroacetamide
Formula: C₂H₄ClNO
MW: 93.51 g/mol
Boiling Pt: 114…116 °C (760 mmHg)
Melting Pt: –75 °C
Density: 0.815 g/cm³
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00008027
CAS Number: 79-07-2
EINECS: 209-526-7
UN: 1105
ADR: 3,III
Merck Index: 13,07197

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4090 to 1.4110 (20°C, 589 nm)
GC ≥97.5 %
Iron (Fe) ≤5 ppm
Ammonium chloride (NH4Cl) ≤0.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR