Glutaraldehyde 25% (w/w) in water
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Glutaral, Glutardialdehyde, 1,5-Pentanedial
11998-0025
119980010
119980100
119980250
TS11998-0025EA
239
USD
TS11998-0025
TS119980010
TS119980100
TS119980250
Glutaraldehyde 25% (w/w) in water
Glutaraldehyde
Formula:
C₅H₈O₂ MW: 100.12 Boiling Pt: 188 Melting Pt: -10 Density: 1.06 g/cm³ Storage Temperature: Refrigerator |
MDL Number:
MFCD00007025 CAS Number: 111-30-8 EINECS: 203-856-5 UN: 3265 ADR: 8 II Merck Index: 13,04485 |
Specification Test Results
Appearance | Clear solution |
Color scale | ≤50 APHA |
Strength | 24 to 26 % |
pH | 3.2 to 4.2 |
Methanol | ≤0.3 % (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...