1-Bromo-2-methoxyethane 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Bromoethyl methyl ether

40298-1000 40298-0050 40298-0250 40298-5000
TS40298-1000EA 197 USD
TS40298-1000 TS40298-0050 TS40298-0250 TS40298-5000
1-Bromo-2-methoxyethane 95%
1-Bromo-2-methoxyethane
Formula: BrH₂CCH₂OCH₃
MW: 138.99 g/mol
Boiling Pt: 110
Density: 1.46 g/cm³
Flash Pt: 28 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000236
CAS Number: 6482-24-2
EINECS: 229-339-4
UN: 1993
ADR: 3 III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4460 to 1.4500 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR