Iodine monobromide 98%

Supplier: Thermo Scientific Chemicals
Danger

20884-2500 AC20884-0500
TS20884-2500EA 261 USD
TS20884-2500 TS20884-0500
Iodine monobromide 98%
Iodine monobromide
Formula: IBr
MW: 206.81 g/mol
Boiling Pt: 116 °C (760 mmHg)
Melting Pt: 42…50 °C
Density: 4.41 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00011353
CAS Number: 7789-33-5
EINECS: 232-159-9
UN: 3260
ADR: 8,II
Merck Index: 13,05038

Order Now

Specification Test Results

Appearance Dark gray to brown-black Crystals or crystalline mass and/or chunks
Titration Iodometric ≥97.5 %
Sulfated ash ≤0.05 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR