Pyridine 99+%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Azabenzene, Azine

13178-0010 13178-0500 131780025 131780250
TS13178-0010EA 138 USD
TS13178-0010 TS13178-0500 TS131780025 TS131780250
Pyridine 99+%, Extra Pure
Pyridine
Formula: C₅H₅N
MW: 79.1 g/mol
Boiling Pt: 115…116 °C (760 mmHg)
Melting Pt: –42 °C
Density: 0.978 g/cm³
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3,II
Merck Index: 13,08060
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9c5934940218027dae0d80a1f98b6d5179c0a8&vid=9f2ef46a26e488d366e82869b3acb96652b5dab1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.5080 to 1.5110 (20°C, 589 nm)
Water ≤0.1 % (Coulometric)
GC ≥99.0 %
Color scale ≤40 APHA
Residue after evaporation ≤0.02 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR