Pyridine 99+% for spectroscopy
Supplier: Thermo Scientific Chemicals
|
Danger
|
38656-0250
38656-2500
386560010
TS38656-0250EA
105
USD
TS38656-0250
TS38656-2500
TS386560010
Pyridine 99+% for spectroscopy
Pyridine
Formula:
C₅H₅N Boiling Pt: 115 °C (1013 hPa) Melting Pt: –42 °C Density: 0.978 g/cm³ Flash Pt: 17 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011732 CAS Number: 110-86-1 EINECS: 203-809-9 UN: 1282 ADR: 3,II Merck Index: 13,08060 |
|
Specification Test Results
Appearance | Clear liquid |
Water | ≤0.05 % |
GC | ≥99.0 % |
UV | at 345 nm A: ≤0.022 |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...