Silver nitrate 99+% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
41935-0025
41935-0500
41935-1000
41935-2500
41935-5000
TS41935-0025EA
4449
USD
TS41935-0025
TS41935-0500
TS41935-1000
TS41935-2500
TS41935-5000
Silver nitrate 99+% ACS
Silver nitrate
Formula:
AgNO₃ Boiling Pt: 444 °C (760 mmHg) Melting Pt: 212 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003414 CAS Number: 7761-88-8 EINECS: 231-853-9 UN: 1493 ADR: 5.1,II REACH: 01-2119513705-43 Merck Index: 12,08661 |
|
Specification Test Results
Appearance | White Crystalline powder or crystals |
Free acid | Passes test |
Titration | ≥99.0 % |
Clarity of solution | Passes test |
Chloride (Cl) | ≤5 ppm |
Sulfate (SO4) | ≤20 ppm |
Copper (Cu) | ≤2 ppm |
Iron (Fe) | ≤2 ppm |
Lead (Pb) | ≤10 ppm |
Subst not pptd by HCl | ≤0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...