Silver nitrate 99.85% for analysis

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

19768-0025 19768-0250 AC19768-0050 AC19768-1000 AC19768-5000
TS19768-0025EA 3975 USD
TS19768-0025 TS19768-0250 TS19768-0050 TS19768-1000 TS19768-5000
Silver nitrate 99.85% for analysis
Silver nitrate
Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: 444
Melting Pt: 210
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 1493
ADR: 5.1 II
REACH: 01-2119513705-43
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Loss on drying ≤0.1 % (1 g, 70°C, 2 h)
Titration with HCl ≥99.80 %
Chloride (Cl) ≤5 ppm
Nitrite (NO2) ≤10 ppm
Cadmium (Cd) ≤1 ppm
Calcium (Ca) ≤10 ppm
Chromium (Cr) ≤1 ppm
Copper (Cu) ≤1 ppm
Iron (Fe) ≤1 ppm
Lead (Pb) ≤1 ppm
Manganese (Mn) ≤1 ppm
Nickel (Ni) ≤1 ppm
Zinc (Zn) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR