Silver nitrate 99.5% for analysis
Supplier: Thermo Scientific Chemicals
|
Danger
|
41936-0250
41936-1000
41936-5000
TS41936-0250EA
154
USD
TS41936-0250
TS41936-1000
TS41936-5000
Silver nitrate 99.5% for analysis
Silver nitrate
Formula:
AgNO₃ Boiling Pt: 444 °C (1013 hPa) Melting Pt: 212 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003414 CAS Number: 7761-88-8 EINECS: 231-853-9 UN: 1493 ADR: 5.1,II REACH: 01-2119513705-43 Merck Index: 12,08661 |
|
Specification Test Results
Appearance | White Crystalline powder or crystals |
Titration Argentometric | ≥99.4 % |
Chloride (Cl) | ≤5 ppm |
Cadmium (Cd) | ≤1 ppm |
Calcium (Ca) | ≤10 ppm |
Chromium (Cr) | ≤0.2 ppm |
Copper (Cu) | ≤0.1 ppm |
Iron (Fe) | ≤0.4 ppm |
Lead (Pb) | ≤0.2 ppm |
Manganese (Mn) | ≤0.1 ppm |
Nickel (Ni) | ≤0.2 ppm |
Palladium (Pd) | ≤0.1 ppm |
Sulfur (S) | ≤20 ppm |
Zinc (Zn) | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...