Phosphorus(III) bromide 99%

Supplier: Thermo Scientific Chemicals
Danger

16947-1000 AC16947-0050 AC16947-5000
TS16947-1000EA 81.68 USD
TS16947-1000 TS16947-0050 TS16947-5000
Phosphorus(III) bromide 99%
Phosphorus(III) bromide
Formula: PBr₃
Boiling Pt: 175 °C (760 mmHg)
Melting Pt: –40 °C
Density: 2.88 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011436
CAS Number: 7789-60-8
EINECS: 232-178-2
UN: 1808
ADR: 8,II
Merck Index: 12,07514

Order Now

Specification Test Results

Appearance Clear to slightly hazy colorless to yellow to light brown Liquid
Titration Argentometric ≥98.5 % (ex Br)
Melting point 110 to 116 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR