Sodium orthovanadate 99%

Supplier: Thermo Scientific Chemicals
Warning

20533-0500 AC20533-2500
TS20533-0500EA 144 USD
TS20533-0500 TS20533-2500
Sodium orthovanadate 99%
Sodium orthovanadate
Formula: Na₃O₄V
Melting Pt: 850…866 °C
Storage Temperature: Ambient
MDL Number: MFCD00003511
CAS Number: 13721-39-6
EINECS: 237-287-9

Order Now

Specification Test Results

Appearance White to off-white Powder
Assay ≥98.5 % (as Na3VO4)
Aluminium (Al) ≤200 ppm
Iron (Fe) ≤30 ppm
Potassium (K) ≤150 ppm
Silicon (Si) ≤200 ppm
Vanadium (V) 26.5 % (typically)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR