Sodium sulfate, anhydrous 99%, Extra Pure

Supplier: Thermo Scientific Chemicals

19664-0251 AC19664-0010 AC19664-0025 AC19664-0050 AC19664-0250
TS19664-0251EA 652 USD
TS19664-0251 TS19664-0010 TS19664-0025 TS19664-0050 TS19664-0250
Sodium sulfate, anhydrous 99%, Extra Pure
Sodium sulphate
Formula: Na₂SO₄
Boiling Pt: 1429 °C (1013 hPa)
Melting Pt: 884 °C
Storage Temperature: Ambient
MDL Number: MFCD00003504
CAS Number: 7757-82-6
EINECS: 231-820-9
Merck Index: 12,08829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
Assay ≥98.5 %
Water ≤0.1 %
Sodium chloride (NaCl) ≤3000 ppm
Iron (Fe) ≤10 ppm
Zinc (Zn) ≤200 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR